COM448626454 Combi-Blocks, Inc.

4-Chloro-2-fluoroanisole

  • MDL number MFCD00672964
  • Cas Number : 452-09-5
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4503

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4503
Availability details for
Quantity
  • Availability
  • Add to Cart