COM448626457 Combi-Blocks, Inc.

4-Chloro-3-fluorophenetole

  • MDL number MFCD00672979
  • Cas Number : 289039-33-4
  • Molecular Weight : 174.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4508

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4508
Availability details for
Quantity
  • Availability
  • Add to Cart