COM448626523 Combi-Blocks, Inc.

4,4'-Dibutoxybiphenyl

  • MDL number MFCD00059419
  • Cas Number : 39800-63-0
  • Molecular Weight : 298.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4646

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4646
Availability details for
Quantity
  • Availability
  • Add to Cart