COM448626565 Combi-Blocks, Inc.

4-Amino-2,2-dimethyl-butyric acid

  • MDL number MFCD01076202
  • Cas Number : 153039-15-7
  • Molecular Weight : 131.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4727

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4727
Availability details for
Quantity
  • Availability
  • Add to Cart