COM448626591 Combi-Blocks, Inc.

3,3-Bis(n,n-dipropanoic acid)

  • MDL number MFCD01311684
  • Cas Number : 505-47-5
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4797

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4797
Availability details for
Quantity
  • Availability
  • Add to Cart