COM448626601 Combi-Blocks, Inc.

L-N-(4'-N-Cbz-piperidino)proline

  • MDL number MFCD01317801
  • Cas Number : 289677-06-1
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4818

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4818
Availability details for
Quantity
  • Availability
  • Add to Cart