COM448626662 Combi-Blocks, Inc.

2-(Aminoethyl)-1-n-boc-piperidine

  • MDL number MFCD01631187
  • Cas Number : 239482-98-5
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4953

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4953
Availability details for
Quantity
  • Availability
  • Add to Cart