COM448626738 Combi-Blocks, Inc.

1-(4-Nitrobenzyl)-1h-imidazole

  • MDL number MFCD01799096
  • Cas Number : 18994-90-6
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5102
Availability details for
Quantity
  • Availability
  • Add to Cart