COM448626748 Combi-Blocks, Inc.

(S)-3-Amino-1-ethylazepan-2-one

  • MDL number MFCD01861209
  • Cas Number : 206434-45-9
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5117

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5117
Availability details for
Quantity
  • Availability
  • Add to Cart