COM448626849 Combi-Blocks, Inc.

5-Fluoro-2-(1h-imidazol-1-yl)aniline

  • MDL number MFCD02180945
  • Cas Number : 251649-52-2
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5266

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5266
Availability details for
Quantity
  • Availability
  • Add to Cart