COM448627050 Combi-Blocks, Inc.

4-Bromo-3-methyl-2-thiophenecarbonyl chloride

  • MDL number MFCD03265447
  • Cas Number : 265652-36-6
  • Molecular Weight : 239.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5638

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5638
Availability details for
Quantity
  • Availability
  • Add to Cart