COM448627063 Combi-Blocks, Inc.

2-Methyl-4-iodo-1-tritylimidazole

  • MDL number MFCD03411213
  • Cas Number : 157255-72-6
  • Molecular Weight : 450.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5664

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5664
Availability details for
Quantity
  • Availability
  • Add to Cart