COM448627079 Combi-Blocks, Inc.

(1-Methyl-3-(4-(trifluoromethyl)phenyl)-1h-pyrazol-5-yl)methanol

  • MDL number MFCD17926173
  • Cas Number : 639784-65-9
  • Molecular Weight : 256.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5689

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5689
Availability details for
Quantity
  • Availability
  • Add to Cart