COM448627108 Combi-Blocks, Inc.

1-Boc-tryptamine

  • MDL number MFCD03426148
  • Cas Number : 167015-84-1
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5757

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5757
Availability details for
Quantity
  • Availability
  • Add to Cart