COM448627116 Combi-Blocks, Inc.

N-Hydroxypropionamidine

  • MDL number MFCD03426272
  • Cas Number : 29335-36-2
  • Molecular Weight : 88.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5807

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5807
Availability details for
Quantity
  • Availability
  • Add to Cart