COM448627118 Combi-Blocks, Inc.

Methyl 1-amino-1-cyclopentanecarboxylate

  • MDL number MFCD03426284
  • Cas Number : 78388-61-1
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5813
Availability details for
Quantity
  • Availability
  • Add to Cart