COM448627140 Combi-Blocks, Inc.

4-Chloro-2-(1-piperidino)-5-thiazolecarboxaldehyde

  • MDL number MFCD03453084
  • Cas Number : 139670-00-1
  • Molecular Weight : 230.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5871

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5871
Availability details for
Quantity
  • Availability
  • Add to Cart