COM448627195 Combi-Blocks, Inc.

2-Methyl-3-(4'-methoxybenzoyl)indole

  • MDL number MFCD03839860
  • Cas Number : 26211-90-5
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5983

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5983
Availability details for
Quantity
  • Availability
  • Add to Cart