COM448627198 Combi-Blocks, Inc.

4-Boc-aminomethylbenzamidine

  • MDL number MFCD03839883
  • Cas Number : 162696-15-3
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5986

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5986
Availability details for
Quantity
  • Availability
  • Add to Cart