COM448627202 Combi-Blocks, Inc.

4-Fluoro-2-methylindole-3-carboxylic acid ethyl ester

  • MDL number MFCD03839918
  • Cas Number : 886362-68-1
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5990

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5990
Availability details for
Quantity
  • Availability
  • Add to Cart