COM448627241 Combi-Blocks, Inc.

(4-Benzyl-morpholin-2-yl)-acetic acid

  • MDL number MFCD04038495
  • Cas Number : 146944-27-6
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6070

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6070
Availability details for
Quantity
  • Availability
  • Add to Cart