COM448627242 Combi-Blocks, Inc.

1-(3-Bromephenyl)-piperazin-2-one hcl

  • MDL number MFCD04038499
  • Cas Number : 215649-81-3
  • Molecular Weight : 291.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6073

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6073
Availability details for
Quantity
  • Availability
  • Add to Cart