COM448627496 Combi-Blocks, Inc.

(1R,2S)-2-(Isopropylamino)-1,2-diphenylethanol

  • MDL number MFCD00798212
  • Cas Number : 71653-81-1
  • Molecular Weight : 255.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6646

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6646
Availability details for
Quantity
  • Availability
  • Add to Cart