COM448627582 Combi-Blocks, Inc.

5-Ethylpyridine-2,3-dicarboxylic acid diethyl ester

  • MDL number MFCD06658212
  • Cas Number : 105151-39-1
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6878

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6878
Availability details for
Quantity
  • Availability
  • Add to Cart