COM448627607 Combi-Blocks, Inc.

2-(4-Bromomethylphenyl)pyridine

  • MDL number MFCD06659494
  • Cas Number : 52199-24-3
  • Molecular Weight : 248.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6916

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6916
Availability details for
Quantity
  • Availability
  • Add to Cart