COM448627632 Combi-Blocks, Inc.

4-Chloro-6-fluoro-quinoline-3-carbonitrile

  • MDL number MFCD06796583
  • Cas Number : 886362-73-8
  • Molecular Weight : 206.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6954

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6954
Availability details for
Quantity
  • Availability
  • Add to Cart