COM448627699 Combi-Blocks, Inc.

2-Amino-4-chloro-6-hydroxy-5-nitropyrimidine

  • MDL number MFCD07367841
  • Cas Number : 1007-99-4
  • Molecular Weight : 190.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7115
Availability details for
Quantity
  • Availability
  • Add to Cart