COM448627709 Combi-Blocks, Inc.

4,5-Diamino-1-(2-hydroxyethyl)pyrazole sulfate

  • MDL number MFCD07368638
  • Cas Number : 155601-30-2
  • Molecular Weight : 240.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7134
Availability details for
Quantity
  • Availability
  • Add to Cart