COM448627710 Combi-Blocks, Inc.

(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanecarboxylic acid

  • MDL number MFCD00798812
  • Cas Number : 16052-40-7
  • Molecular Weight : 184.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7135

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7135
Availability details for
Quantity
  • Availability
  • Add to Cart