COM448627820 Combi-Blocks, Inc.

(S)-3-Cbz-amino-2-piperidone

  • MDL number MFCD08166408
  • Cas Number : 95582-17-5
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7544

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7544
Availability details for
Quantity
  • Availability
  • Add to Cart