COM448627923 Combi-Blocks, Inc.

2-Amino-3-cyano-4-methyl-5-iodopyridine

  • MDL number MFCD09030796
  • Cas Number : 180995-02-2
  • Molecular Weight : 259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8204

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8204
Availability details for
Quantity
  • Availability
  • Add to Cart