COM448627942 Combi-Blocks, Inc.

1-Propylpyrrolidin-2-one

  • MDL number MFCD09749878
  • Cas Number : 3470-99-3
  • Molecular Weight : 127.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8252

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8252
Availability details for
Quantity
  • Availability
  • Add to Cart