COM448627970 Combi-Blocks, Inc.

2,5-Difluoro-3-methylbenzoic acid

  • MDL number MFCD09835286
  • Cas Number : 1003710-01-7
  • Molecular Weight : 172.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8355

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8355
Availability details for
Quantity
  • Availability
  • Add to Cart