COM448628015 Combi-Blocks, Inc.

(2-Amino-2-methyl-propyl)-urea

  • MDL number MFCD03839821
  • Cas Number : 87484-83-1
  • Molecular Weight : 131.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8490
Availability details for
Quantity
  • Availability
  • Add to Cart