COM448628036 Combi-Blocks, Inc.

(2R)-Methyl 2-methyl-5-oxocyclopentanecarboxylate

  • MDL number MFCD11502470
  • Cas Number : 92344-02-0
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8529

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8529
Availability details for
Quantity
  • Availability
  • Add to Cart