COM448628038 Combi-Blocks, Inc.

(2R,4S)-3-Benzoyl-4-ethyl-4-methyl-2-phenyloxazolidin-5-one

  • MDL number MFCD07784040
  • Cas Number : 1279025-92-1
  • Molecular Weight : 309.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8534

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8534
Availability details for
Quantity
  • Availability
  • Add to Cart