COM448628052 Combi-Blocks, Inc.

(2S,3S)-2-Amino-3-hydroxy-4-methyl-valeric acid

  • MDL number MFCD09842579
  • Cas Number : 10148-70-6
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8563

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8563
Availability details for
Quantity
  • Availability
  • Add to Cart