COM448628056 Combi-Blocks, Inc.

(2S,4S)-1-Boc-4-benzylpyrrolidine-2-dicarboxylic acid ethyl ester

  • MDL number MFCD18711395
  • Cas Number : 170947-82-7
  • Molecular Weight : 333.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8577

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8577
Availability details for
Quantity
  • Availability
  • Add to Cart