COM448628059 Combi-Blocks, Inc.

(2S,4S)-1-Isopropyl-4-methoxy-proline

  • MDL number MFCD11053655
  • Cas Number : 1820571-30-9
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8580

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8580
Availability details for
Quantity
  • Availability
  • Add to Cart