COM448628065 Combi-Blocks, Inc.

(2S,4S)-4-tert-Butoxy-1-isopropyl-proline

  • MDL number MFCD11053549
  • Cas Number : 1263078-26-7
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8586

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8586
Availability details for
Quantity
  • Availability
  • Add to Cart