COM448628071 Combi-Blocks, Inc.

(3,4-Difluoro-phenyl)-piperidin-4-yl-methanone

  • MDL number MFCD03839837
  • Cas Number : 149452-43-7
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8598

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8598
Availability details for
Quantity
  • Availability
  • Add to Cart