COM448628073 Combi-Blocks, Inc.

(3,5-Bis(trifluoromethyl)phenyl)(phenyl)methanol

  • MDL number MFCD11100929
  • Cas Number : 81390-93-4
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8601

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8601
Availability details for
Quantity
  • Availability
  • Add to Cart