COM448628076 Combi-Blocks, Inc.

(3',5'-Difluoro-biphenyl-3-yl)-acetic acid

  • MDL number MFCD03839973
  • Cas Number : 866108-77-2
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8605

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8605
Availability details for
Quantity
  • Availability
  • Add to Cart