COM448628077 Combi-Blocks, Inc.

(3-[4-(Trifluoromethyl)phenoxy]phenyl)sulfonyl chloride

  • MDL number MFCD02089490
  • Cas Number : 882014-01-9
  • Molecular Weight : 336.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8609
Availability details for
Quantity
  • Availability
  • Add to Cart