COM448628082 Combi-Blocks, Inc.

(3-Bromobenzyl)-cyanoacetic acid

  • MDL number MFCD09800398
  • Cas Number : 948015-61-0
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8617

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8617
Availability details for
Quantity
  • Availability
  • Add to Cart