COM448628089 Combi-Blocks, Inc.

(3'-Ethoxy-biphenyl-4-yl)-acetic acid

  • MDL number MFCD03426477
  • Cas Number : 669713-69-3
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8629

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8629
Availability details for
Quantity
  • Availability
  • Add to Cart