COM448628101 Combi-Blocks, Inc.

(3R)-3-Amino-3-(3-bromophenyl)propan-1-ol

  • MDL number MFCD07784054
  • Cas Number : 1213827-47-4
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8648

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8648
Availability details for
Quantity
  • Availability
  • Add to Cart