COM448628108 Combi-Blocks, Inc.

(3S,4R)-1-Boc-3-hydroxymethyl-4-(4-fluorophenyl)-piperidine

  • MDL number MFCD10687161
  • Cas Number : 200572-33-4
  • Molecular Weight : 309.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8667

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8667
Availability details for
Quantity
  • Availability
  • Add to Cart