COM448628114 Combi-Blocks, Inc.

(4-(Chloromethyl)phenyl)methanamine hcl

  • MDL number MFCD09954377
  • Cas Number : 1194752-34-5
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8685

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8685
Availability details for
Quantity
  • Availability
  • Add to Cart