COM448628119 Combi-Blocks, Inc.

(4'-Chloro-biphenyl-2-yl)acetic acid

  • MDL number MFCD03426506
  • Cas Number : 669713-87-5
  • Molecular Weight : 246.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8700
Availability details for
Quantity
  • Availability
  • Add to Cart